Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATTTGGGCTAAAAGACACACTA[C/T]CTTAAAAGTTTCACATTCTTTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103060 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADSS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADSS - adenylosuccinate synthase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C1orf101 - chromosome 1 open reading frame 101 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130957.1 | 150 | Intron | NP_001124429.1 | |||
NM_001242340.1 | 150 | Intron | NP_001229269.1 | |||
NM_173807.4 | 150 | Intron | NP_776168.1 | |||
XM_011544138.2 | 150 | Intron | XP_011542440.1 | |||
XM_011544139.2 | 150 | Intron | XP_011542441.1 | |||
XM_011544142.2 | 150 | Intron | XP_011542444.1 | |||
XM_011544144.2 | 150 | Intron | XP_011542446.1 | |||
XM_011544145.2 | 150 | Intron | XP_011542447.1 | |||
XM_011544149.2 | 150 | Intron | XP_011542451.1 | |||
XM_011544151.2 | 150 | Intron | XP_011542453.1 | |||
XM_017000943.1 | 150 | Intron | XP_016856432.1 | |||
XM_017000944.1 | 150 | UTR 5 | XP_016856433.1 | |||
XM_017000945.1 | 150 | Intron | XP_016856434.1 | |||
XM_017000946.1 | 150 | Intron | XP_016856435.1 | |||
XM_017000947.1 | 150 | Intron | XP_016856436.1 | |||
XM_017000948.1 | 150 | Intron | XP_016856437.1 | |||
XM_017000949.1 | 150 | Intron | XP_016856438.1 | |||
XM_017000950.1 | 150 | Intron | XP_016856439.1 | |||
XM_017000951.1 | 150 | Intron | XP_016856440.1 | |||
XM_017000952.1 | 150 | Intron | XP_016856441.1 | |||
XM_017000953.1 | 150 | Intron | XP_016856442.1 |