Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGCCGTTCTGTTACCAAAGAAA[A/G]CCGAAAGCCACAAAGCCAAAAGCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 615014 MIM: 602797 MIM: 601831 | ||||||||||||||||||||
Literature Links: |
HIST2H2AB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIST2H2AB - histone cluster 2, H2ab | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175065.2 | 361 | Intron | NP_778235.1 |
HIST2H2AC - histone cluster 2, H2ac | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003517.2 | 361 | Missense Mutation | ACC,GCC | T,A 121 | NP_003508.1 |
HIST2H2BE - histone cluster 2, H2be | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003528.2 | 361 | Intron | NP_003519.1 |