Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCATCTGAATCACTGTCCTCCAT[C/G]ATGGCGGCAGGCTGGTGGTAGAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606015 MIM: 604136 | ||||||||||||||||||||
Literature Links: |
FCMR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FCMR - Fc fragment of IgM receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142473.1 | 1248 | Missense Mutation | ATC,ATG | I,M 264 | NP_001135945.1 | |
NM_001193338.1 | 1248 | UTR 3 | NP_001180267.1 | |||
NM_005449.4 | 1248 | Missense Mutation | ATC,ATG | I,M 376 | NP_005440.1 | |
XM_005273351.3 | 1248 | UTR 3 | XP_005273408.1 | |||
XM_005273352.4 | 1248 | Missense Mutation | ATC,ATG | I,M 285 | XP_005273409.1 |
IL24 - interleukin 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185156.1 | 1248 | Intron | NP_001172085.1 | |||
NM_001185157.1 | 1248 | Intron | NP_001172086.1 | |||
NM_001185158.1 | 1248 | Intron | NP_001172087.1 | |||
NM_006850.3 | 1248 | Intron | NP_006841.1 | |||
XM_011509101.2 | 1248 | Intron | XP_011507403.1 | |||
XM_017000121.1 | 1248 | Intron | XP_016855610.1 |