Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTCATCAGACTGTGGCTACACA[A/G]TCTCTCAAGTGAGATCAATGAAGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608003 MIM: 610088 | ||||||||||||||||||||
Literature Links: |
HIPK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIPK1 - homeodomain interacting protein kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OLFML3 - olfactomedin like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286352.1 | 693 | Missense Mutation | ATC,GTC | I,V 119 | NP_001273281.1 | |
NM_001286353.1 | 693 | Missense Mutation | ATC,GTC | I,V 78 | NP_001273282.1 | |
NM_020190.3 | 693 | Missense Mutation | ATC,GTC | I,V 139 | NP_064575.1 | |
XM_017001848.1 | 693 | Missense Mutation | ATC,GTC | I,V 119 | XP_016857337.1 |