Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAACCTCACCGACTTCTTGATG[G/T]AAATGGGCTTCCGCATGGACCATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612384 | ||||||||||||||||||||
Literature Links: |
MED18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MED18 - mediator complex subunit 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127350.1 | Intron | NP_001120822.1 | ||||
NM_017638.2 | Intron | NP_060108.2 | ||||
XM_005245914.4 | Intron | XP_005245971.1 |