Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCACAGAGGGCAGGGTGGTGACT[A/G]AGGACCAGCTCCTCATGCTTGAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
THEMIS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
THEMIS2 - thymocyte selection associated family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039477.2 | 405 | Intron | NP_001034566.1 | |||
NM_001105556.2 | 405 | Missense Mutation | AAG,GAG | K,E 126 | NP_001099026.1 | |
NM_001286113.1 | 405 | Missense Mutation | AAG,GAG | K,E 126 | NP_001273042.1 | |
NM_001286115.1 | 405 | Missense Mutation | AAG,GAG | K,E 126 | NP_001273044.1 | |
NM_004848.3 | 405 | Missense Mutation | AAG,GAG | K,E 126 | NP_004839.2 | |
XM_005246041.3 | 405 | Intron | XP_005246098.1 | |||
XM_006711050.1 | 405 | Missense Mutation | AAG,GAG | K,E 126 | XP_006711113.1 | |
XM_011542445.2 | 405 | Missense Mutation | AAG,GAG | K,E 79 | XP_011540747.1 |