Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACATAGGTAGTGGATAAAGGTGC[C/T]AGCATATGACACACTCCATTTGCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600682 | ||||||||||||||||||||
Literature Links: |
AKR7A2P1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKR7A2P1 - aldo-keto reductase family 7 member A2 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC16A1 - solute carrier family 16 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166496.1 | 1401 | Silent Mutation | CTA,CTG | L,L 346 | NP_001159968.1 | |
NM_003051.3 | 1401 | Silent Mutation | CTA,CTG | L,L 346 | NP_003042.3 | |
XM_011542026.2 | 1401 | Intron | XP_011540328.1 | |||
XM_011542027.2 | 1401 | Intron | XP_011540329.1 |