Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGCTCATGGGCAAGGGCTGGGC[C/T]AAGGGGCTCCTTCCCGAAGTCCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604188 | ||||||||||||||||||||
Literature Links: |
SELENBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SELENBP1 - selenium binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258288.1 | 1656 | Missense Mutation | AGC,GGC | S,G 390 | NP_001245217.1 | |
NM_001258289.1 | 1656 | Missense Mutation | AGC,GGC | S,G 494 | NP_001245218.1 | |
NM_003944.3 | 1656 | Missense Mutation | AGC,GGC | S,G 452 | NP_003935.2 | |
XM_017002757.1 | 1656 | Intron | XP_016858246.1 |