Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAACAATACGATGGTGATAATAC[C/T]AGTGTTAGAGGGATGCTGTGGGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164175 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POU2F1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POU2F1 - POU class 2 homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198783.1 | Intron | NP_001185712.1 | ||||
NM_001198786.1 | Intron | NP_001185715.1 | ||||
NM_002697.3 | Intron | NP_002688.3 | ||||
XM_011509653.2 | Intron | XP_011507955.1 | ||||
XM_011509654.2 | Intron | XP_011507956.1 | ||||
XM_011509655.1 | Intron | XP_011507957.1 | ||||
XM_017001508.1 | Intron | XP_016856997.1 |