Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGACAGCTACGGAGGCCCAAGC[A/C]CCCCAGGGCCAGCACAAGCTCCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SERTAD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SERTAD4 - SERTA domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019605.3 | 494 | Missense Mutation | ACC,CCC | T,P 39 | NP_062551.1 | |
XM_005273193.4 | 494 | Missense Mutation | ACC,CCC | T,P 39 | XP_005273250.1 |
SERTAD4-AS1 - SERTAD4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |