Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTAGAGGCTTGGTGGTCCTGACC[C/T]CAGAGAGAAGCTATACCCTGGAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605548 MIM: 601380 | ||||||||||||||||||||
Literature Links: |
ADAM15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAM15 - ADAM metallopeptidase domain 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261464.1 | 554 | Missense Mutation | CCA,TCA | P,S 157 | NP_001248393.1 | |
NM_001261465.1 | 554 | Missense Mutation | CCA,TCA | P,S 147 | NP_001248394.1 | |
NM_001261466.1 | 554 | Missense Mutation | CCA,TCA | P,S 131 | NP_001248395.1 | |
NM_003815.4 | 554 | Missense Mutation | CCA,TCA | P,S 147 | NP_003806.3 | |
NM_207191.2 | 554 | Missense Mutation | CCA,TCA | P,S 147 | NP_997074.1 | |
NM_207194.2 | 554 | Missense Mutation | CCA,TCA | P,S 147 | NP_997077.1 | |
NM_207195.2 | 554 | Missense Mutation | CCA,TCA | P,S 147 | NP_997078.1 | |
NM_207196.2 | 554 | Missense Mutation | CCA,TCA | P,S 147 | NP_997079.1 | |
NM_207197.2 | 554 | Missense Mutation | CCA,TCA | P,S 147 | NP_997080.1 |
DCST1 - DC-STAMP domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EFNA4 - ephrin A4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100505666 - uncharacterized LOC100505666 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |