Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTGCTGGTGGCGCGGGGAGTGC[A/G]CGTCAAGGTGAACGAGGCCTACCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600347 | ||||||||||||||||||||
Literature Links: |
BCAN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCAN - brevican | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021948.4 | 626 | Missense Mutation | CAC,CGC | H,R 97 | NP_068767.3 | |
NM_198427.1 | 626 | Missense Mutation | CAC,CGC | H,R 97 | NP_940819.1 | |
XM_011509866.1 | 626 | Missense Mutation | CAC,CGC | H,R 97 | XP_011508168.1 | |
XM_017002047.1 | 626 | Missense Mutation | CAC,CGC | H,R 142 | XP_016857536.1 |