Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGAATGGAAAGAAGGGCTGCCT[A/G]CAAGAGCTCTTCAGAAAATTCAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600236 | ||||||||||||||||||||
Literature Links: |
CENPF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CENPF - centromere protein F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016343.3 | 201 | Missense Mutation | ACA,GCA | T,A 14 | NP_057427.3 | |
XM_011509082.2 | 201 | Missense Mutation | ACA,GCA | T,A 14 | XP_011507384.1 | |
XM_017000086.1 | 201 | Missense Mutation | ACA,GCA | T,A 14 | XP_016855575.1 |