Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATCTTCTTTGTGAACCCTGCACC[C/T]CCATACCTGTGGCCAGCCCACAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 103320 MIM: 147571 | ||||||||||||||||||||
Literature Links: |
AGRN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AGRN - agrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305275.1 | 434 | Silent Mutation | CCC,CCT | P,P 121 | NP_001292204.1 | |
NM_198576.3 | 434 | Silent Mutation | CCC,CCT | P,P 121 | NP_940978.2 | |
XM_005244749.3 | 434 | Silent Mutation | CCC,CCT | P,P 121 | XP_005244806.1 | |
XM_011541429.2 | 434 | Silent Mutation | CCC,CCT | P,P 121 | XP_011539731.1 |
ISG15 - ISG15 ubiquitin-like modifier | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |