Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCATCTCTGGAGTTTGACTGTAG[G/T]CCTGGTGGACCTGGCCCCTCAGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102775 MIM: 160795 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADORA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADORA1 - adenosine A1 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000674.2 | 1559 | Intron | NP_000665.1 | |||
NM_001048230.1 | 1559 | Intron | NP_001041695.1 | |||
XM_005244901.1 | 1559 | Intron | XP_005244958.1 | |||
XM_005244902.4 | 1559 | Intron | XP_005244959.1 | |||
XM_017000342.1 | 1559 | Intron | XP_016855831.1 |
MYBPH - myosin binding protein H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004997.2 | 1559 | UTR 3 | NP_004988.2 |