Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGCACTGTTAAGGTCAGATAAG[C/T]ACCTGCCCTGGGCATAACTGAAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603031 | ||||||||||||||||||||
Literature Links: |
TLR5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TLR5 - toll like receptor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003268.5 | 2829 | Missense Mutation | TAC,TGC | Y,C 779 | NP_003259.2 | |
XM_005273241.4 | 2829 | Missense Mutation | TAC,TGC | Y,C 779 | XP_005273298.2 | |
XM_005273242.4 | 2829 | Missense Mutation | TAC,TGC | Y,C 779 | XP_005273299.2 | |
XM_005273243.4 | 2829 | Missense Mutation | TAC,TGC | Y,C 779 | XP_005273300.2 | |
XM_006711504.3 | 2829 | Missense Mutation | TAC,TGC | Y,C 779 | XP_006711567.1 | |
XM_006711505.3 | 2829 | Missense Mutation | TAC,TGC | Y,C 779 | XP_006711568.1 | |
XM_006711506.3 | 2829 | Missense Mutation | TAC,TGC | Y,C 779 | XP_006711569.1 | |
XM_011509937.2 | 2829 | Missense Mutation | TAC,TGC | Y,C 779 | XP_011508239.1 | |
XM_017002208.1 | 2829 | Missense Mutation | TAC,TGC | Y,C 779 | XP_016857697.1 |