Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATCCCTGACATTGATGCGAATGA[A/C]GCCATCCATTCTGGGCCAGAAGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602758 MIM: 610568 | ||||||||||||||||||||
Literature Links: |
LOC100507670 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100507670 - uncharacterized LOC100507670 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PI4KB - phosphatidylinositol 4-kinase beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF687 - zinc finger protein 687 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304763.1 | 377 | Missense Mutation | GAA,GAC | E,D 25 | NP_001291692.1 | |
NM_001304764.1 | 377 | Missense Mutation | GAA,GAC | E,D 25 | NP_001291693.1 | |
NM_020832.2 | 377 | Missense Mutation | GAA,GAC | E,D 25 | NP_065883.1 | |
XM_005245366.3 | 377 | Missense Mutation | GAA,GAC | E,D 34 | XP_005245423.1 | |
XM_011509811.2 | 377 | Missense Mutation | GAA,GAC | E,D 25 | XP_011508113.1 | |
XM_011509812.2 | 377 | Missense Mutation | GAA,GAC | E,D 25 | XP_011508114.1 | |
XM_011509813.2 | 377 | Missense Mutation | GAA,GAC | E,D 25 | XP_011508115.1 |