Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGCCGCGAGTGTCCCTTTCCTT[C/T]CTGGCGCTTTCCTGGGTTGCTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120920 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD46 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD46 - CD46 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002389.4 | 213 | Missense Mutation | TCC,TTC | S,F 13 | NP_002380.3 | |
NM_153826.3 | 213 | Missense Mutation | TCC,TTC | S,F 13 | NP_722548.1 | |
NM_172350.2 | 213 | Missense Mutation | TCC,TTC | S,F 13 | NP_758860.1 | |
NM_172351.2 | 213 | Missense Mutation | TCC,TTC | S,F 13 | NP_758861.1 | |
NM_172352.2 | 213 | Missense Mutation | TCC,TTC | S,F 13 | NP_758862.1 | |
NM_172353.2 | 213 | Missense Mutation | TCC,TTC | S,F 13 | NP_758863.1 | |
NM_172359.2 | 213 | Missense Mutation | TCC,TTC | S,F 13 | NP_758869.1 | |
NM_172361.2 | 213 | Missense Mutation | TCC,TTC | S,F 13 | NP_758871.1 | |
XM_011509563.1 | 213 | Missense Mutation | TCC,TTC | S,F 13 | XP_011507865.1 | |
XM_011509564.1 | 213 | Missense Mutation | TCC,TTC | S,F 13 | XP_011507866.1 | |
XM_017001308.1 | 213 | Missense Mutation | TCC,TTC | S,F 13 | XP_016856797.1 | |
XM_017001309.1 | 213 | Missense Mutation | TCC,TTC | S,F 13 | XP_016856798.1 | |
XM_017001310.1 | 213 | Missense Mutation | TCC,TTC | S,F 13 | XP_016856799.1 |