Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCTGGGCTCCAATTAGTAATGC[C/T]TGGAGTGATGCCCAAGTACATCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610955 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAPPC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRAPPC3 - trafficking protein particle complex 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270894.1 | 212 | Missense Mutation | AGC,GGC | S,G 100 | NP_001257823.1 | |
NM_001270895.1 | 212 | Missense Mutation | AGC,GGC | S,G 46 | NP_001257824.1 | |
NM_001270896.1 | 212 | Missense Mutation | AGC,GGC | S,G 46 | NP_001257825.1 | |
NM_001270897.1 | 212 | Missense Mutation | AGC,GGC | S,G 26 | NP_001257826.1 | |
NM_014408.4 | 212 | Missense Mutation | AGC,GGC | S,G 92 | NP_055223.1 |