Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGCCAGGACCCGGCGTGCCAGGC[A/G]TGCCTCCACCATGCGTTCTATGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616136 | ||||||||||||||||||||
Literature Links: |
RNF220 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF220 - ring finger protein 220 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM53 - transmembrane protein 53 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300746.1 | 1049 | Missense Mutation | CGC,TGC | R,C 162 | NP_001287675.1 | |
NM_001300747.1 | 1049 | Missense Mutation | CGC,TGC | R,C 205 | NP_001287676.1 | |
NM_001300748.1 | 1049 | Missense Mutation | CGC,TGC | R,C 204 | NP_001287677.1 | |
NM_024587.3 | 1049 | Missense Mutation | CGC,TGC | R,C 235 | NP_078863.2 | |
XM_011542138.2 | 1049 | Missense Mutation | CGC,TGC | R,C 118 | XP_011540440.1 |