Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAGAAACCAAGTATTTGAGCCC[A/G]GCTGCCTGGATGCCTTCCCAAACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 138380 MIM: 138333 | ||||||||||||||||||||
Literature Links: |
GSTM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSTM2 - glutathione S-transferase mu 2 (muscle) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000848.3 | 611 | Missense Mutation | AGC,GGC | S,G 173 | NP_000839.1 | |
NM_001142368.1 | 611 | Missense Mutation | AGC,GGC | S,G 173 | NP_001135840.1 |
GSTM4 - glutathione S-transferase mu 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |