Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCATTCAGCCGCTGTAGAGCCC[A/G]CTGCACCAGGGCCTGGTACTCCCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604468 MIM: 608042 | ||||||||||||||||||||
Literature Links: |
MAP3K6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP3K6 - mitogen-activated protein kinase kinase kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297609.1 | 3822 | Missense Mutation | CGG,TGG | R,W 1185 | NP_001284538.1 | |
NM_004672.4 | 3822 | Missense Mutation | CGG,TGG | R,W 1193 | NP_004663.3 | |
XM_017002771.1 | 3822 | Missense Mutation | CGG,TGG | R,W 774 | XP_016858260.1 | |
XM_017002772.1 | 3822 | Intron | XP_016858261.1 |
SYTL1 - synaptotagmin like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |