Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTCACTGTCTTCAAGAAGCTCCT[A/G]AACCCCAAATGTCGCGATGTCTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610624 MIM: 120252 MIM: 608953 | ||||||||||||||||||||
Literature Links: |
ADPRHL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADPRHL2 - ADP-ribosylhydrolase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017825.2 | 437 | Silent Mutation | CTA,CTG | L,L 128 | NP_060295.1 | |
XM_011541636.2 | 437 | UTR 5 | XP_011539938.1 |
COL8A2 - collagen type VIII alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEKT2 - tektin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |