Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTATTCTAGATCCACTCAGTACA[C/T]CAGCGCCGGGAAGTCCAGGGCCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ATXN7L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATXN7L2 - ataxin 7 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153340.4 | 813 | Silent Mutation | CAC,CAT | H,H 266 | NP_699171.3 | |
XM_005270445.3 | 813 | Silent Mutation | CAC,CAT | H,H 298 | XP_005270502.1 | |
XM_005270446.2 | 813 | Silent Mutation | CAC,CAT | H,H 298 | XP_005270503.1 | |
XM_011540638.2 | 813 | Silent Mutation | CAC,CAT | H,H 276 | XP_011538940.1 | |
XM_011540639.2 | 813 | Silent Mutation | CAC,CAT | H,H 240 | XP_011538941.1 | |
XM_011540641.2 | 813 | Silent Mutation | CAC,CAT | H,H 240 | XP_011538943.1 | |
XM_011540644.2 | 813 | Silent Mutation | CAC,CAT | H,H 240 | XP_011538946.1 | |
XM_011540645.1 | 813 | Silent Mutation | CAC,CAT | H,H 83 | XP_011538947.1 | |
XM_011540646.2 | 813 | Silent Mutation | CAC,CAT | H,H 298 | XP_011538948.1 | |
XM_017000267.1 | 813 | Silent Mutation | CAC,CAT | H,H 240 | XP_016855756.1 | |
XM_017000268.1 | 813 | Silent Mutation | CAC,CAT | H,H 240 | XP_016855757.1 | |
XM_017000269.1 | 813 | Silent Mutation | CAC,CAT | H,H 240 | XP_016855758.1 | |
XM_017000270.1 | 813 | Silent Mutation | CAC,CAT | H,H 240 | XP_016855759.1 |
CYB561D1 - cytochrome b561 family member D1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYPL2 - synaptophysin like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |