Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAGAAGAATTAGAGAACTATAGG[C/T]CCTCTGAGCAGAGGAAACAGCATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120650 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CR2 - complement component 3d receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006658.2 | Intron | NP_001006659.1 | ||||
NM_001877.4 | Intron | NP_001868.2 | ||||
XM_011509206.2 | Intron | XP_011507508.1 |