Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCACCACAGGGAGGACTGTTTC[A/G]TATAAAAAGGATGCATTCTGTCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 615283 MIM: 616086 | ||||||||||||||||||||
Literature Links: |
EXOC8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EXOC8 - exocyst complex component 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175876.4 | 2173 | Silent Mutation | TAC,TAT | Y,Y 682 | NP_787072.2 |
SPRTN - SprT-like N-terminal domain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |