Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCCGTCTTGACCTACACTTGGG[C/T]GCACACCGGCAGCGATGCCGCACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF672 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF672 - zinc finger protein 672 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024836.2 | 1118 | Silent Mutation | GGC,GGT | G,G 90 | NP_079112.1 | |
XM_005270336.1 | 1118 | Silent Mutation | GGC,GGT | G,G 90 | XP_005270393.1 |
ZNF692 - zinc finger protein 692 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |