Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATGTTTGTTGAATGAATGAATG[A/G]CAAGCACTTGTCACAACGGTAACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
STRIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STRIP1 - striatin interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270768.1 | Intron | NP_001257697.1 | ||||
NM_033088.3 | Intron | NP_149079.2 | ||||
XM_006710995.2 | Intron | XP_006711058.1 | ||||
XM_017002629.1 | Intron | XP_016858118.1 | ||||
XM_017002630.1 | Intron | XP_016858119.1 |