Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGGTTATCCGAGAAGTTCCGA[G/T]GATTCTCCTTGGATTTCCTGGGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609738 MIM: 604634 | ||||||||||||||||||||
Literature Links: |
IGSF9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGSF9 - immunoglobulin superfamily member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAGLN2 - transgelin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277223.1 | 786 | Intron | NP_001264152.1 | |||
NM_001277224.1 | 786 | Missense Mutation | CAT,CCT | H,P 180 | NP_001264153.1 | |
NM_003564.2 | 786 | Intron | NP_003555.1 |