Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACAGTGAATTTGCTGGACACGTC[A/G]ACCATCCACGGGGACTGGGGCTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 176945 | ||||||||||||||||||||
Literature Links: |
EPHA8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPHA8 - EPH receptor A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011540969.1 | 136 | UTR 5 | XP_011539271.1 | |||
XM_011540970.1 | 136 | UTR 5 | XP_011539272.1 | |||
XM_011540972.1 | 136 | UTR 5 | XP_011539274.1 | |||
XM_011540973.1 | 136 | Intron | XP_011539275.1 | |||
XM_011540975.2 | 136 | Intron | XP_011539277.1 |