Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGTTGCGAGTGGCGGCTTCTCTC[A/G]TGTGCACCAGGACTGTGAAGGTACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605647 MIM: 604094 | ||||||||||||||||||||
Literature Links: |
FBXO6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO6 - F-box protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAD2L2 - MAD2 mitotic arrest deficient-like 2 (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127325.1 | 504 | Missense Mutation | ACG,ATG | T,M 152 | NP_001120797.1 | |
NM_006341.3 | 504 | Missense Mutation | ACG,ATG | T,M 152 | NP_006332.3 | |
XM_011540507.1 | 504 | Missense Mutation | ACG,ATG | T,M 152 | XP_011538809.1 |