Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAAGCAACTCCAACTTTGATTTC[C/T]GTAAACATGTCCAAAATATCCAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 103060 | ||||||||||||||||||||
Literature Links: |
ADSS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADSS - adenylosuccinate synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126.3 | 2099 | Silent Mutation | ACA,ACG | T,T 370 | NP_001117.2 | |
XM_011544108.2 | 2099 | Silent Mutation | ACA,ACG | T,T 310 | XP_011542410.1 |