Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATATTGTTTCCAGACAGAGTCAAG[C/T]GGAACATCCTGGCCATGGGCGAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 147892 | ||||||||||||||||||||
Literature Links: |
DIO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DIO1 - deiodinase, iodothyronine type I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000792.6 | 139 | Missense Mutation | CGG,TGG | R,W 39 | NP_000783.2 | |
NM_001039715.2 | 139 | Missense Mutation | CGG,TGG | R,W 39 | NP_001034804.1 | |
NM_001039716.2 | 139 | Missense Mutation | CGG,TGG | R,W 39 | NP_001034805.1 | |
NM_001324316.1 | 139 | Missense Mutation | CGG,TGG | R,W 39 | NP_001311245.1 | |
NM_213593.4 | 139 | Missense Mutation | CGG,TGG | R,W 39 | NP_998758.1 |
YIPF1 - Yip1 domain family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |