Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGGAATAAACATCAAAGTTCTCC[A/C]GCTCTTTGCGGCTCAATCGAGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605554 MIM: 600684 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD244 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD244 - CD244 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166663.1 | 1313 | Missense Mutation | CGG,CTG | R,L 363 | NP_001160135.1 | |
NM_001166664.1 | 1313 | Missense Mutation | CGG,CTG | R,L 266 | NP_001160136.1 | |
NM_016382.3 | 1313 | Missense Mutation | CGG,CTG | R,L 358 | NP_057466.1 | |
XM_011509621.2 | 1313 | Intron | XP_011507923.1 | |||
XM_011509622.2 | 1313 | Intron | XP_011507924.1 | |||
XM_011509623.2 | 1313 | Missense Mutation | CGG,CTG | R,L 160 | XP_011507925.1 |
LY9 - lymphocyte antigen 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |