Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGAGTGGCCTCTCCTCCATCAAA[C/T]GCTGTGTGGATGAGGACCTGGAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 136131 | ||||||||||||||||||||
Literature Links: |
FMO4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FMO4 - flavin containing monooxygenase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002022.2 | 491 | Missense Mutation | CGC,TGC | R,C 20 | NP_002013.1 | |
XM_005245045.3 | 491 | Intron | XP_005245102.1 | |||
XM_005245046.3 | 491 | Missense Mutation | CGC,TGC | R,C 20 | XP_005245103.1 | |
XM_005245048.3 | 491 | Missense Mutation | CGC,TGC | R,C 20 | XP_005245105.1 | |
XM_006711243.3 | 491 | Missense Mutation | CGC,TGC | R,C 20 | XP_006711306.1 | |
XM_011509347.2 | 491 | Missense Mutation | CGC,TGC | R,C 20 | XP_011507649.1 | |
XM_011509348.2 | 491 | Intron | XP_011507650.1 | |||
XM_011509349.2 | 491 | Missense Mutation | CGC,TGC | R,C 20 | XP_011507651.1 |