Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGAAGCAGTTAACCGAATACCC[C/G]CTTGACATACCCCTGAACACCCGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615218 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC400794 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC400794 - uncharacterized LOC400794 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC52 - leucine rich repeat containing 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005214.3 | 200 | Silent Mutation | CCC,CCG | P,P 48 | NP_001005214.2 |