Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTGGTGTACCTGTCTCAGTTGA[C/T]AAGGACGTGCATATTCCTTTCACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616136 | ||||||||||||||||||||
Literature Links: |
RNF220 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF220 - ring finger protein 220 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319956.1 | 821 | Silent Mutation | GAC,GAT | D,D 55 | NP_001306885.1 | |
NM_001319957.1 | 821 | Intron | NP_001306886.1 | |||
NM_018150.3 | 821 | Silent Mutation | GAC,GAT | D,D 55 | NP_060620.2 | |
XM_005270992.1 | 821 | Silent Mutation | GAC,GAT | D,D 55 | XP_005271049.1 | |
XM_005270993.1 | 821 | Silent Mutation | GAC,GAT | D,D 55 | XP_005271050.1 | |
XM_005270996.1 | 821 | Intron | XP_005271053.1 | |||
XM_006710735.3 | 821 | Silent Mutation | GAC,GAT | D,D 55 | XP_006710798.1 | |
XM_011541698.1 | 821 | Silent Mutation | GAC,GAT | D,D 55 | XP_011540000.1 | |
XM_011541699.1 | 821 | Silent Mutation | GAC,GAT | D,D 55 | XP_011540001.1 | |
XM_011541700.1 | 821 | Silent Mutation | GAC,GAT | D,D 55 | XP_011540002.1 | |
XM_011541702.1 | 821 | Intron | XP_011540004.1 | |||
XM_017001623.1 | 821 | Silent Mutation | GAC,GAT | D,D 55 | XP_016857112.1 | |
XM_017001624.1 | 821 | Silent Mutation | GAC,GAT | D,D 55 | XP_016857113.1 | |
XM_017001625.1 | 821 | Silent Mutation | GAC,GAT | D,D 55 | XP_016857114.1 | |
XM_017001626.1 | 821 | Intron | XP_016857115.1 |