Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACCACCCGGACCTGCTCCACCA[A/C]CAGAAGGGAGGGCCCGTCCTTTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 615242 | ||||||||||||||||||||
Literature Links: |
CCDC27 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC27 - coiled-coil domain containing 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC47 - leucine rich repeat containing 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020710.2 | 1673 | Missense Mutation | GTG,TTG | V,L 549 | NP_065761.1 |
SMIM1 - small integral membrane protein 1 (Vel blood group) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |