Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGAAAGGCCTCTGGCCGGTGTGC[A/G]GCGTCTGGTGCTCGGACAGATTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF648 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF648 - zinc finger protein 648 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009992.1 | 5463 | Missense Mutation | CCG,CTG | P,L 441 | NP_001009992.1 | |
XM_017000301.1 | 5463 | Missense Mutation | CCG,CTG | P,L 441 | XP_016855790.1 | |
XM_017000302.1 | 5463 | Missense Mutation | CCG,CTG | P,L 441 | XP_016855791.1 |