Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCGAATGGCTTGGGGATGCCCA[C/G]GTCCTTGTCCAGCACGATCATGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607935 | ||||||||||||||||||||
Literature Links: |
PADI2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PADI2 - peptidyl arginine deiminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007365.2 | 1048 | Missense Mutation | CTG,GTG | L,V 598 | NP_031391.2 | |
XM_017000148.1 | 1048 | Missense Mutation | CTG,GTG | L,V 283 | XP_016855637.1 |