Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATGCTCAATTTTCATTGCCAA[C/T]GTTCTTGGTAGATTCACTCTTCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNHIT6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNHIT6 - zinc finger HIT-type containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170670.1 | 1545 | Missense Mutation | ATT,GTT | I,V 427 | NP_001164141.1 | |
NM_017953.3 | 1545 | Missense Mutation | ATT,GTT | I,V 466 | NP_060423.3 | |
XM_011541614.2 | 1545 | Intron | XP_011539916.1 |