Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACCAGCCCATCTGGCCCACTGT[C/G]GTGGCCTTGACAAAGCCCTCAGCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607945 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADIPOR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADIPOR1 - adiponectin receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290553.1 | 1191 | Intron | NP_001277482.1 | |||
NM_001290557.1 | 1191 | Silent Mutation | ACC,ACG | T,T 295 | NP_001277486.1 | |
NM_001290629.1 | 1191 | Intron | NP_001277558.1 | |||
NM_015999.5 | 1191 | Silent Mutation | ACC,ACG | T,T 295 | NP_057083.2 | |
XM_017001407.1 | 1191 | Intron | XP_016856896.1 |