Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTAGTAGAAATCAAATAACCAG[C/G]ACTTACTAGATAATGGTGCCACATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605743 MIM: 603305 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HHAT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HHAT - hedgehog acyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNH1 - potassium voltage-gated channel subfamily H member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002238.3 | 6082 | UTR 3 | NP_002229.1 | |||
NM_172362.2 | 6082 | UTR 3 | NP_758872.1 | |||
XM_017001246.1 | 6082 | UTR 3 | XP_016856735.1 |