Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGCCATGCAGGAGAAATATCCA[A/G]CTGAGGGGATCTCTCACGTCACTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CEP85 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP85 - centrosomal protein 85 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281517.2 | 205 | Missense Mutation | ACT,GCT | T,A 9 | NP_001268446.1 | |
NM_001281518.2 | 205 | Intron | NP_001268447.1 | |||
NM_001319944.1 | 205 | Missense Mutation | ACT,GCT | T,A 9 | NP_001306873.1 | |
NM_022778.4 | 205 | Missense Mutation | ACT,GCT | T,A 9 | NP_073615.2 | |
XM_017002105.1 | 205 | Missense Mutation | ACT,GCT | T,A 9 | XP_016857594.1 |
LOC101928303 - uncharacterized LOC101928303 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |