Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGCCAAAGCATTGCTGCTCACCT[A/G]GATAACCAGGTTCCAGTTGAGAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602074 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAP3 - death associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199849.1 | 284 | Silent Mutation | CTA,CTG | L,L 36 | NP_001186778.1 | |
NM_001199850.1 | 284 | Silent Mutation | CTA,CTG | L,L 36 | NP_001186779.1 | |
NM_001199851.1 | 284 | Intron | NP_001186780.1 | |||
NM_004632.3 | 284 | Silent Mutation | CTA,CTG | L,L 36 | NP_004623.1 | |
NM_033657.2 | 284 | Silent Mutation | CTA,CTG | L,L 36 | NP_387506.1 | |
XM_005245480.2 | 284 | Silent Mutation | CTA,CTG | L,L 36 | XP_005245537.1 | |
XM_005245481.2 | 284 | Intron | XP_005245538.1 | |||
XM_005245482.4 | 284 | Intron | XP_005245539.1 | |||
XM_011509972.2 | 284 | Silent Mutation | CTA,CTG | L,L 36 | XP_011508274.1 | |
XM_017002289.1 | 284 | Silent Mutation | CTA,CTG | L,L 36 | XP_016857778.1 | |
XM_017002290.1 | 284 | Silent Mutation | CTA,CTG | L,L 36 | XP_016857779.1 | |
XM_017002291.1 | 284 | Silent Mutation | CTA,CTG | L,L 36 | XP_016857780.1 | |
XM_017002292.1 | 284 | Intron | XP_016857781.1 | |||
XM_017002293.1 | 284 | Intron | XP_016857782.1 | |||
XM_017002294.1 | 284 | Silent Mutation | CTA,CTG | L,L 36 | XP_016857783.1 | |
XM_017002295.1 | 284 | Intron | XP_016857784.1 | |||
XM_017002296.1 | 284 | Intron | XP_016857785.1 | |||
XM_017002297.1 | 284 | Intron | XP_016857786.1 |