Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGGTTGGCACTGCTTCTGGTTC[C/T]GCTGGCAGGACATCTTGGCAGGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612617 | ||||||||||||||||||||
Literature Links: |
LCE3E PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LCE3E - late cornified envelope 3E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178435.3 | 51 | Missense Mutation | CAG,CGG | Q,R 5 | NP_848522.1 | |
XM_011509484.1 | 51 | Missense Mutation | CAG,CGG | Q,R 5 | XP_011507786.1 |