Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGGTGACGGCAAACAAGGCCCA[C/T]ACTGGACAGGGCAGCTGCTGGGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602135 MIM: 609365 MIM: 608241 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNALI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNALI1 - dynein axonemal light intermediate chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003462.3 | 16 | Silent Mutation | CAC,CAT | H,H 8 | NP_003453.2 | |
XM_005271172.2 | 16 | UTR 5 | XP_005271229.1 |
GNL2 - G protein nucleolar 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNIP1 - Smad nuclear interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |