Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATTTCATTTCCTTCCAGCATTG[C/T]CTAATGAAATAGCTGGCACTTTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602208 MIM: 610273 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNJ10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNJ10 - potassium voltage-gated channel subfamily J member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002241.4 | 2638 | UTR 3 | NP_002232.2 |
PIGM - phosphatidylinositol glycan anchor biosynthesis class M | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |