Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCAGTTGAGTACCAGTGCCAGAA[C/T]TTGTATCAACTTGAGGGTAACAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600889 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFHR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFHR2 - complement factor H related 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001312672.1 | 472 | Silent Mutation | AAC,AAT | N,N 56 | NP_001299601.1 | |
NM_005666.3 | 472 | Silent Mutation | AAC,AAT | N,N 180 | NP_005657.1 | |
XM_005245113.3 | 472 | Silent Mutation | AAC,AAT | N,N 115 | XP_005245170.1 | |
XM_011509458.2 | 472 | Silent Mutation | AAC,AAT | N,N 176 | XP_011507760.1 | |
XM_011509459.2 | 472 | Silent Mutation | AAC,AAT | N,N 160 | XP_011507761.1 | |
XM_011509460.2 | 472 | Silent Mutation | AAC,AAT | N,N 111 | XP_011507762.1 | |
XM_017001109.1 | 472 | Silent Mutation | AAC,AAT | N,N 121 | XP_016856598.1 |