Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTCTTTTCTAAATTACCTTGTA[C/G]TCTTGTCCAGATTTACTCTGCAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611350 | ||||||||||||||||||||
Literature Links: |
INTS7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INTS7 - integrator complex subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199809.1 | 2776 | Silent Mutation | GAC,GAG | D,E 816 | NP_001186738.1 | |
NM_001199811.1 | 2776 | Silent Mutation | GAC,GAG | D,E 851 | NP_001186740.1 | |
NM_001199812.1 | 2776 | Silent Mutation | GAC,GAG | D,E 845 | NP_001186741.1 | |
NM_015434.3 | 2776 | Silent Mutation | GAC,GAG | D,E 865 | NP_056249.1 | |
XM_011509396.2 | 2776 | Silent Mutation | GAC,GAG | D,E 901 | XP_011507698.1 | |
XM_017000962.1 | 2776 | Intron | XP_016856451.1 |